Learn More
Benzyl 3-bromopropyl ether, 95%, Thermo Scientific Chemicals
$67.74 - $224.62
Chemical Identifiers
CAS | 54314-84-0 |
---|---|
Molecular Formula | C10H13BrO |
Molecular Weight (g/mol) | 229.12 |
MDL Number | MFCD00134570 |
InChI Key | PSUXTZLDBVEZTD-UHFFFAOYSA-N |
Synonym | benzyl 3-bromopropyl ether, 3-bromopropoxy methyl benzene, 3-bromopropylbenzyl ether, 3-benzyloxy-1-bromopropane, 3-benzyloxy propyl bromide, benzene, 3-bromopropoxy methyl, 1-3-bromopropoxy methyl benzene, benzyl3-bromopropylether |
PubChem CID | 2776064 |
IUPAC Name | [(3-bromopropoxy)methyl]benzene |
SMILES | BrCCCOCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH6627803
|
Thermo Scientific Chemicals
H6627803 |
1 g |
Each for $67.74
|
|
||||||
AAH6627806
|
Thermo Scientific Chemicals
H6627806 |
5 g |
Each for $224.62
|
|
||||||
Description
Benzyl 3-bromopropyl ether may be used in the studies of preparation of (2S,3S)-1-[(triisopropylsilyl)oxy]-7-(benzyloxy)-2,3-(isopropylidenedioxy)-4(Z)-heptene, Ppeparation of 5-(3-Benzyloxypropoxy)psoralen (PAP-7), total synthesis of zincophorin and (+)-anatoxin-a.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzyl 3-bromopropyl ether may be used in the studies of preparation of (2S,3S)-1-[(triisopropylsilyl)oxy]-7-(benzyloxy)-2,3-(isopropylidenedioxy)-4(Z)-heptene, Ppeparation of 5-(3-Benzyloxypropoxy)psoralen (PAP-7), total synthesis of zincophorin and (+)-anatoxin-a.
Solubility
Slightly soluble in water.
Notes
Store under dry inert gas. Store away from air. Incompatible with air and oxidizing agents.
Chemical Identifiers
54314-84-0 | |
229.12 | |
PSUXTZLDBVEZTD-UHFFFAOYSA-N | |
2776064 | |
BrCCCOCC1=CC=CC=C1 |
C10H13BrO | |
MFCD00134570 | |
benzyl 3-bromopropyl ether, 3-bromopropoxy methyl benzene, 3-bromopropylbenzyl ether, 3-benzyloxy-1-bromopropane, 3-benzyloxy propyl bromide, benzene, 3-bromopropoxy methyl, 1-3-bromopropoxy methyl benzene, benzyl3-bromopropylether | |
[(3-bromopropoxy)methyl]benzene |
Specifications
54314-84-0 | |
1.298 g/mL | |
113°C (235°F) | |
1.531 | |
1 g | |
benzyl 3-bromopropyl ether, 3-bromopropoxy methyl benzene, 3-bromopropylbenzyl ether, 3-benzyloxy-1-bromopropane, 3-benzyloxy propyl bromide, benzene, 3-bromopropoxy methyl, 1-3-bromopropoxy methyl benzene, benzyl3-bromopropylether | |
PSUXTZLDBVEZTD-UHFFFAOYSA-N | |
[(3-bromopropoxy)methyl]benzene | |
2776064 | |
95% | |
Benzyl 3-bromopropyl ether |
Colorless | |
130°C to 132°C (8 mmHg) | |
C10H13BrO | |
MFCD00134570 | |
2084412 | |
Slightly soluble in water. | |
BrCCCOCC1=CC=CC=C1 | |
229.12 | |
229.12 | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only