missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzyl 4-aminopiperidine-1-carboxylate, 97%, Thermo Scientific™
$693.75 - $693.75
Chemical Identifiers
CAS | 120278-07-1 |
---|---|
Molecular Formula | C13H18N2O2 |
Molecular Weight (g/mol) | 234.299 |
MDL Number | MFCD05863884 |
InChI Key | YYIQGSYCCNQAGV-UHFFFAOYSA-N |
Synonym | 1-n-cbz-4-aminopiperidine, 4-amino-1-n-cbz-piperidine, 1-cbz-4-aminopiperidine, 4-amino-piperidine-1-carboxylic acid benzyl ester, 4-amino-1-cbz-piperidine, 1-n-cbz-4-amino-piperidine, 1-piperidinecarboxylic acid, 4-amino-, phenylmethyl ester, 4-aminopiperidine-1-carboxylic acid benzyl ester, benzyl 4-aminopiperidine-1-carboxylate; 4-aminopiperidine-1-carboxylic acid benzyl ester, pubchem19775 |
PubChem CID | 17846640 |
IUPAC Name | benzyl 4-aminopiperidine-1-carboxylate |
SMILES | C1CN(CCC1N)C(=O)OCC2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6442406
|
Thermo Scientific Chemicals
H6442406 |
5 g |
Each for $693.75
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
120278-07-1 | |
234.299 | |
YYIQGSYCCNQAGV-UHFFFAOYSA-N | |
17846640 | |
C1CN(CCC1N)C(=O)OCC2=CC=CC=C2 |
C13H18N2O2 | |
MFCD05863884 | |
1-n-cbz-4-aminopiperidine, 4-amino-1-n-cbz-piperidine, 1-cbz-4-aminopiperidine, 4-amino-piperidine-1-carboxylic acid benzyl ester, 4-amino-1-cbz-piperidine, 1-n-cbz-4-amino-piperidine, 1-piperidinecarboxylic acid, 4-amino-, phenylmethyl ester, 4-aminopiperidine-1-carboxylic acid benzyl ester, benzyl 4-aminopiperidine-1-carboxylate; 4-aminopiperidine-1-carboxylic acid benzyl ester, pubchem19775 | |
benzyl 4-aminopiperidine-1-carboxylate |
Specifications
120278-07-1 | |
MFCD05863884 | |
10003356 | |
Slightly soluble in water. | |
C1CN(CCC1N)C(=O)OCC2=CC=CC=C2 | |
234.299 | |
234.3 | |
Benzyl 4-aminopiperidine-1-carboxylate |
C13H18N2O2 | |
5 g | |
1-n-cbz-4-aminopiperidine, 4-amino-1-n-cbz-piperidine, 1-cbz-4-aminopiperidine, 4-amino-piperidine-1-carboxylic acid benzyl ester, 4-amino-1-cbz-piperidine, 1-n-cbz-4-amino-piperidine, 1-piperidinecarboxylic acid, 4-amino-, phenylmethyl ester, 4-aminopiperidine-1-carboxylic acid benzyl ester, benzyl 4-aminopiperidine-1-carboxylate; 4-aminopiperidine-1-carboxylic acid benzyl ester, pubchem19775 | |
YYIQGSYCCNQAGV-UHFFFAOYSA-N | |
benzyl 4-aminopiperidine-1-carboxylate | |
17846640 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures