missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzyl 4-chlorophenyl ketone, 98%, Thermo Scientific Chemicals
$87.99 - $1,070.34
Chemical Identifiers
CAS | 1889-71-0 |
---|---|
Molecular Formula | C14H11ClO |
Molecular Weight (g/mol) | 230.69 |
MDL Number | MFCD00016342 |
InChI Key | DXVALSKCLLBZEB-UHFFFAOYSA-N |
Synonym | benzyl 4-chlorophenyl ketone, 1-4-chlorophenyl-2-phenylethanone, 4'-chloro-2-phenylacetophenone, 4-chlorodeoxybenzoin, 1-4-chlorophenyl-2-phenylethan-1-one, 1-4-chlorophenyl-2-phenyl-ethanone, 4-chlorodesoxy benzoin, chlorodeoxybenzoin, deoxy-4-chlorobenzoin, acmc-20a6by |
PubChem CID | 233840 |
IUPAC Name | 1-(4-chlorophenyl)-2-phenylethanone |
SMILES | ClC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1595906
|
Thermo Scientific Chemicals
A1595906 |
5 g |
N/A
|
|
|||||
AAA1595914
|
Thermo Scientific Chemicals
A1595914 |
25 g |
N/A
|
|
|||||
AAA1595922
|
Thermo Scientific Chemicals
A1595922 |
100 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1889-71-0 | |
230.69 | |
DXVALSKCLLBZEB-UHFFFAOYSA-N | |
233840 | |
ClC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1 |
C14H11ClO | |
MFCD00016342 | |
benzyl 4-chlorophenyl ketone, 1-4-chlorophenyl-2-phenylethanone, 4'-chloro-2-phenylacetophenone, 4-chlorodeoxybenzoin, 1-4-chlorophenyl-2-phenylethan-1-one, 1-4-chlorophenyl-2-phenyl-ethanone, 4-chlorodesoxy benzoin, chlorodeoxybenzoin, deoxy-4-chlorobenzoin, acmc-20a6by | |
1-(4-chlorophenyl)-2-phenylethanone |
Specifications
1889-71-0 | |
C14H11ClO | |
5 g | |
benzyl 4-chlorophenyl ketone, 1-4-chlorophenyl-2-phenylethanone, 4'-chloro-2-phenylacetophenone, 4-chlorodeoxybenzoin, 1-4-chlorophenyl-2-phenylethan-1-one, 1-4-chlorophenyl-2-phenyl-ethanone, 4-chlorodesoxy benzoin, chlorodeoxybenzoin, deoxy-4-chlorobenzoin, acmc-20a6by | |
ClC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1 | |
230.69 | |
230.69 | |
Benzyl 4-chlorophenyl ketone |
103°C to 107°C | |
MFCD00016342 | |
976443 | |
DXVALSKCLLBZEB-UHFFFAOYSA-N | |
1-(4-chlorophenyl)-2-phenylethanone | |
233840 | |
98% |
Safety and Handling
P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P501c
H317-H318
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only