missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-Hydroxybenzoate 98.0+%, TCI America™

$42.87 - $44.73

Chemical Identifiers

CAS 94-18-8
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD00016471
InChI Key MOZDKDIOPSPTBH-UHFFFAOYSA-N
Synonym benzylparaben, benzyl p-hydroxybenzoate, nipabenzyl, parosept, benzyl paraben, benzyl parasept, benzyl tegosept, solbrol z, benzyl-parasept, p-hydroxybenzoic acid benzyl ester
PubChem CID 7180
ChEBI CHEBI:34571
IUPAC Name benzyl 4-hydroxybenzoate
SMILES OC1=CC=C(C=C1)C(=O)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B37685G
View Documents
TCI America
B37685G
5 g
Each for $42.87
Only null left
 
B376825G
View Documents
TCI America
B376825G
25 g
Each for $44.73
Only null left
 
Description

Description

Specifications

Chemical Identifiers

94-18-8
228.25
MOZDKDIOPSPTBH-UHFFFAOYSA-N
7180
benzyl 4-hydroxybenzoate
C14H12O3
MFCD00016471
benzylparaben, benzyl p-hydroxybenzoate, nipabenzyl, parosept, benzyl paraben, benzyl parasept, benzyl tegosept, solbrol z, benzyl-parasept, p-hydroxybenzoic acid benzyl ester
CHEBI:34571
OC1=CC=C(C=C1)C(=O)OCC1=CC=CC=C1

Specifications

94-18-8
White
MFCD00016471
3077
MOZDKDIOPSPTBH-UHFFFAOYSA-N
benzyl 4-hydroxybenzoate
7180
228.25
Crystalline Powder
113°C
C14H12O3
5 g
benzylparaben, benzyl p-hydroxybenzoate, nipabenzyl, parosept, benzyl paraben, benzyl parasept, benzyl tegosept, solbrol z, benzyl-parasept, p-hydroxybenzoic acid benzyl ester
OC1=CC=C(C=C1)C(=O)OCC1=CC=CC=C1
228.25
CHEBI:34571
≥98.0% (GC,T)
Benzyl 4-Hydroxybenzoate [for Biochemical Research]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1325

RTECSNumber : DH2691000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'