Learn More
Benzyl Alcohol, 99%, Pure
CAS: 100-51-6 | C7H8O | 108.14 g/mol
$165.40 - $1703.74
Chemical Identifiers
| CAS | 100-51-6 |
|---|---|
| Molecular Formula | C7H8O |
| Molecular Weight (g/mol) | 108.14 |
| MDL Number | MFCD00004599,MFCD03792087 |
| InChI Key | WVDDGKGOMKODPV-UHFFFAOYSA-N |
| Synonym | benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol |
| PubChem CID | 244 |
| ChEBI | CHEBI:17987 |
| IUPAC Name | phenylmethanol |
| SMILES | OCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC148390010
|
Thermo Scientific Chemicals
148390010 |
1 L | Glass bottle |
Each for $165.40
Case of 4 Each for $0.00
|
|
||||
|
AC148390250
|
Thermo Scientific Chemicals
148390250 |
25 L | Metal drum |
Each for $1,703.74
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 100-51-6 | |
| 108.14 | |
| WVDDGKGOMKODPV-UHFFFAOYSA-N | |
| 244 | |
| phenylmethanol |
| C7H8O | |
| MFCD00004599,MFCD03792087 | |
| benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
| CHEBI:17987 | |
| OCC1=CC=CC=C1 |
Specifications
| 100-51-6 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| 1.5380 to 1.5410 (20°C, 589nm) | |
| MFCD00004599,MFCD03792087 | |
| 06, 428 | |
| benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
| OCC1=CC=CC=C1 | |
| 108.14 | |
| CHEBI:17987 | |
| 0.13mbar at 20°C | |
| Pure | |
| Benzyl alcohol |
| 98.5 | |
| Colorless to Yellow | |
| 99% | |
| C7H8O | |
| C6H5CH2OH | |
| 1 L | |
| 15, 1127 | |
| WVDDGKGOMKODPV-UHFFFAOYSA-N | |
| phenylmethanol | |
| 244 | |
| 108.14 | |
| ≥98.5% | |
| Liquid |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/phys
GHS Signal Word: Warning
EINECSNumber : 202-859-9
RUO – Research Use Only