Learn More
Benzyl alcohol, for analysis
CAS: 100-51-6 | C7H8O | 108.14 g/mol
$309.88 - $584.67
Chemical Identifiers
| CAS | 100-51-6 |
|---|---|
| Molecular Formula | C7H8O |
| Molecular Weight (g/mol) | 108.14 |
| MDL Number | MFCD00004599,MFCD03792087 |
| InChI Key | WVDDGKGOMKODPV-UHFFFAOYSA-N |
| Synonym | benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol |
| PubChem CID | 244 |
| ChEBI | CHEBI:17987 |
| IUPAC Name | phenylmethanol |
| SMILES | OCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC391220010
|
Thermo Scientific Chemicals
391220010 |
1 L | Glass bottle |
Each for $309.88
|
|
||||
|
AC391220025
|
Thermo Scientific Chemicals
391220025 |
2.5 L | Glass bottle |
Each for $584.67
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisChemical Identifiers
| 100-51-6 | |
| 108.14 | |
| WVDDGKGOMKODPV-UHFFFAOYSA-N | |
| 244 | |
| phenylmethanol |
| C7H8O | |
| MFCD00004599,MFCD03792087 | |
| benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
| CHEBI:17987 | |
| OCC1=CC=CC=C1 |
Specifications
| 0.002 meq/g max. | |
| 0.001 meq/g max. | |
| 1.0440g/mL | |
| 96°C | |
| Glass bottle | |
| 1.5380 to 1.5410 | |
| MFCD00004599,MFCD03792087 | |
| 1.044 | |
| benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
| WVDDGKGOMKODPV-UHFFFAOYSA-N | |
| phenylmethanol | |
| 244 | |
| CHEBI:17987 | |
| ≥99.5% | |
| Liquid |
| 100-51-6 | |
| -15.0°C | |
| 205.0°C | |
| Authentic | |
| C7H8O | |
| C6H5CH2OH | |
| 1 L | |
| 15, 1127 | |
| Solubility in water: 40g/L (20°C). Other solubilities: miscible with alcohol,ether,acetone,chloroform,,dmf and methylene chloride | |
| OCC1=CC=CC=C1 | |
| 108.14 | |
| 6.6 mPa.s (20°C) | |
| 108.14 | |
| Analysis | |
| Benzyl alcohol |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/phys
GHS Signal Word: Warning
EINECSNumber : 202-859-9
RUO – Research Use Only