missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzyl Alcohol (Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific BPA3964
Specifications
Benzyl Alcohol | |
4°C max. | |
Yellow | |
205°C | |
0.005% max. | |
C7H8O | |
4 L | |
WVDDGKGOMKODPV-UHFFFAOYSA-N | |
phenylmethanol | |
244 | |
108.14 | |
Certified |
100-51-6 | |
-15°C | |
3.72 (Air = 1.0) | |
96°C | |
Glass Bottle | |
MFCD00004599,MFCD03792087 | |
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
OCC1=CC=CC=C1 | |
108.14 | |
CHEBI:17987 | |
0.13mbar at 20°C | |
Liquid |
Chemical Identifiers
100-51-6 | |
108.14 | |
WVDDGKGOMKODPV-UHFFFAOYSA-N | |
244 | |
phenylmethanol |
C7H8O | |
MFCD00004599,MFCD03792087 | |
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol | |
CHEBI:17987 | |
OCC1=CC=CC=C1 |
Safety and Handling
WARNING!
Emergency Overview
Combustible liquid. Irritating to eyes and skin. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.
NFPA
Health:1
Flammability:1
Instability:0