missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl Alcohol (Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific BPA396500

 View more versions of this product

Catalog No. A396-500



Explore available promotions

Description

Description

Specifications

Specifications

Benzyl Alcohol
4°C max.
Yellow
205°C
0.005% max.
C7H8O
500 mL
WVDDGKGOMKODPV-UHFFFAOYSA-N
phenylmethanol
244
108.14
Certified
100-51-6
-15°C
3.72 (Air = 1.0)
96°C
Glass Bottle
MFCD00004599,MFCD03792087
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
OCC1=CC=CC=C1
108.14
CHEBI:17987
0.13mbar at 20°C
Liquid

Chemical Identifiers

100-51-6
108.14
WVDDGKGOMKODPV-UHFFFAOYSA-N
244
phenylmethanol
C7H8O
MFCD00004599,MFCD03792087
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
CHEBI:17987
OCC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Combustible liquid. Irritating to eyes and skin. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:1
Instability:0

SDS Documents

Documents

Promotions

Promotions