missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, Technical, Spectrum™ Chemical

$350.60 - $29582.47

Chemical Identifiers

CAS 100-51-6
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18609054
View Documents
Spectrum Chemical Mfg Cor
B1092500MLGL
500 mL Amber Glass Bottle
Each for $350.60
Only null left
 
18609058
View Documents
Spectrum Chemical Mfg Cor
B10924LTGL
4 L Amber Glass Bottle
Each for $1,250.37
Only null left
 
18609057
View Documents
Spectrum Chemical Mfg Cor
B109220LTMT
20 L Amber Glass Bottle
Each for $3,913.27
Only null left
 
18609055
View Documents
Spectrum Chemical Mfg Cor
B109255GLBL
55 gal. Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

100-51-6

Specifications

100-51-6
205.3°C
500 mL
Technical
1
Amber Glass Bottle
1.042 - 1.047
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.