Learn More
Benzyl azide, 94%, Thermo Scientific Chemicals
$111.58 - $1,281.97
Chemical Identifiers
CAS | 622-79-7 |
---|---|
Molecular Formula | C7H7N3 |
Molecular Weight (g/mol) | 133.154 |
MDL Number | MFCD00013836 |
InChI Key | UDLLFLQFQMACJB-UHFFFAOYSA-N |
Synonym | azidomethyl benzene, benzyl azide, benzylazide, benzene, azidomethyl, alpha-azidotoluene, toluene, .alpha.-azido, unii-hfd57z7j9j, .alpha.-azidotoluene, ccris 8029 |
PubChem CID | 12152 |
IUPAC Name | azidomethylbenzene |
SMILES | C1=CC=C(C=C1)CN=[N+]=[N-] |
Description
Benzyl azide is used in the synthesis of 1,2,3-triazole derivatives. It serves as a reagent used to introduce a PhCH2N group into a molecule.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
622-79-7 | |
133.154 | |
UDLLFLQFQMACJB-UHFFFAOYSA-N | |
12152 | |
C1=CC=C(C=C1)CN=[N+]=[N-] |
C7H7N3 | |
MFCD00013836 | |
azidomethyl benzene, benzyl azide, benzylazide, benzene, azidomethyl, alpha-azidotoluene, toluene, .alpha.-azido, unii-hfd57z7j9j, .alpha.-azidotoluene, ccris 8029 | |
azidomethylbenzene |
Specifications
622-79-7 | |
82°C to 85°C (16 mm) | |
94% | |
1.5341 | |
2 g | |
Insoluble in waterMiscible with ethanol and diethyl ether. Immiscible with water. | |
C1=CC=C(C=C1)CN=[N+]=[N-] | |
133.154 | |
133.15 | |
Benzyl azide |
1.0655 | |
43°C (110°F) | |
C7H7N3 | |
MFCD00013836 | |
azidomethyl benzene, benzyl azide, benzylazide, benzene, azidomethyl, alpha-azidotoluene, toluene, .alpha.-azido, unii-hfd57z7j9j, .alpha.-azidotoluene, ccris 8029 | |
UDLLFLQFQMACJB-UHFFFAOYSA-N | |
azidomethylbenzene | |
12152 | |
Liquid |
Safety and Handling
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Hazard Class: 3; Packaging Group: III
RTECSNumber : XS6650000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only