missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl benzoate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 105860051

 View more versions of this product

Catalog No. AC105860051


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzyl benzoate
0.3mg KOH/g max.
100.0
Colorless
323.0°C
Authentic
Plastic drum
1.5670 to 1.5700
MFCD00003075
09, 121
15, 1130
Solubility in water: 15.3mg/l (20°C). Other solubilities: soluble in alcohol,chloroform and ether,insoluble in glycerol
O=C(OCC1=CC=CC=C1)C1=CC=CC=C1
212.25
10.9 mPa.s (25°C)
212.25
Liquid
120-51-4
99.0
18.0°C to 20.0°C
1.1180g/mL
147°C
99% min. (GC)
C14H12O2
C6H5CO2CH2C6H5
5 L
1.118
ascabiol, benylate, novoscabin, benzoic acid, phenylmethyl ester, scabitox, scobenol, ascabin, benzyl phenylformate, benzoic acid, benzyl ester, phenylmethyl benzoate
SESFRYSPDFLNCH-UHFFFAOYSA-N
benzyl benzoate
2345
CHEBI:41237
99+%

Chemical Identifiers

120-51-4
212.25
SESFRYSPDFLNCH-UHFFFAOYSA-N
2345
benzyl benzoate
C14H12O2
MFCD00003075
ascabiol, benylate, novoscabin, benzoic acid, phenylmethyl ester, scabitox, scobenol, ascabin, benzyl phenylformate, benzoic acid, benzyl ester, phenylmethyl benzoate
CHEBI:41237
O=C(OCC1=CC=CC=C1)C1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Very toxic to aquatic life.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Avoid release to the environment.

GHS Signal Word: Warning

EINECSNumber : 204-402-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only