missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzyl Benzoate, USP, 99-100.5%, Spectrum™ Chemical
BE159, 120-51-4, C14H12O2
Supplier: Spectrum Chemical Mfg Cor BE159200LTBL
Description
Spectrum™ Chemical Benzyl Benzoate, USP is used as a antiparasitic insecticide against human scabies. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
120-51-4 | |
323°C | |
C14H12O2 | |
MFCD00003075 | |
SESFRYSPDFLNCH-UHFFFAOYSA-N | |
benzyl benzoate | |
99 to 100.5% | |
Liquid |
100% | |
Amber Glass Bottle | |
1.568-1.570 | |
200 L | |
O=C(OCC1=CC=CC=C1)C1=CC=CC=C1 | |
212.25 | |
USP |
Chemical Identifiers
120-51-4 | |
212.25 | |
SESFRYSPDFLNCH-UHFFFAOYSA-N | |
O=C(OCC1=CC=CC=C1)C1=CC=CC=C1 |
C14H12O2 | |
MFCD00003075 | |
benzyl benzoate |