missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl bromide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 105875000

 View more versions of this product

Catalog No. AC105875000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Specifications

Benzyl bromide
100-39-0
100.0
Colorless to Yellow
198.0°C to 199.0°C
Authentic
Glass bottle
1.5742 to 1.5762
MFCD00000172
05, 306
1.43
benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
AGEZXYOZHKGVCM-UHFFFAOYSA-N
bromomethylbenzene
7498
171.04
Liquid
98%
97.5
-2°C
1.4300g/mL
86°C
97.5% min. (GC)
C7H7Br
C6H5CH2Br
500 mL
05,25; 13,51
15, 1131
Solubility in water: 436 mg/l (25°C). Other solubilities: slowly decomposes, soluble in alcohol, benzene and ether
C1=CC=C(C=C1)CBr
171.04
CHEBI:59858
98%

Chemical Identifiers

100-39-0
AGEZXYOZHKGVCM-UHFFFAOYSA-N
7498
bromomethylbenzene
MFCD00000172
benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
CHEBI:59858
C1=CC=C(C=C1)CBr
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF

GHS Signal Word: Warning

EINECSNumber : 202-847-3

RTECSNumber : XS7965000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only