missing translation for 'onlineSavingsMsg'
Learn More

Benzyl ether, 99%

CAS: 103-50-4 | C14H14O | 198.26 g/mol

$95.18 - $211.27

Chemical Identifiers

CAS 103-50-4
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.26
MDL Number MFCD00004780
InChI Key MHDVGSVTJDSBDK-UHFFFAOYSA-N
Synonym dibenzyl ether, benzyl ether, benzyl oxide, dibenzylether, oxybis methylene dibenzene, plastikator ba, ba plasticizer, benzylether, ether, dibenzyl, plasticator ba
PubChem CID 7657
ChEBI CHEBI:87411
IUPAC Name phenylmethoxymethylbenzene
SMILES C1=CC=C(C=C1)COCC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC148400010
SDS View Documents
Thermo Scientific Chemicals
148400010
1 L Glass bottle
Each for $95.18
Only null left
 
AC148400025
SDS View Documents
Thermo Scientific Chemicals
148400025
2.5 L Glass bottle
Each for $211.27
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

103-50-4
198.26
MHDVGSVTJDSBDK-UHFFFAOYSA-N
7657
phenylmethoxymethylbenzene
C14H14O
MFCD00004780
dibenzyl ether, benzyl ether, benzyl oxide, dibenzylether, oxybis methylene dibenzene, plastikator ba, ba plasticizer, benzylether, ether, dibenzyl, plasticator ba
CHEBI:87411
C1=CC=C(C=C1)COCC2=CC=CC=C2

Specifications

103-50-4
100.0
Colorless to Yellow
298.0°C
Authentic
Glass bottle
1.5610 to 1.5630
MFCD00004780
06, 434
15, 1135
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,acetone and chloroform
C1=CC=C(C=C1)COCC2=CC=CC=C2
198.26
CHEBI:87411
99%
Benzyl ether, 99%
98.5
1.5°C to 3.5°C
1.0400g/mL
135°C
99%
C14H14O
(C6H5CH2)2O
1 L
1.04
dibenzyl ether, benzyl ether, benzyl oxide, dibenzylether, oxybis methylene dibenzene, plastikator ba, ba plasticizer, benzylether, ether, dibenzyl, plasticator ba
MHDVGSVTJDSBDK-UHFFFAOYSA-N
phenylmethoxymethylbenzene
7657
198.26
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 203-118-2

RTECSNumber : DQ6125000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only