missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl isocyanate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0263314

 View more versions of this product

Catalog No. AAL0263314


Explore available promotions

Description

Description

Benzyl isocyanate, is used as a protected ammonia equivalent in the stereo selective ring-opening of chiral 2,3-epoxy alcohols (from Sharpless asymmetric epoxidation). The key step in the sequence involves intramolecular cyclization of the carbamate intermediate. It is also used in a multicomponent, stereospecific synthesis of 1,3-oxazinane-2,4-diones employing an Al-salph complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl isocyanate, is used as a protected ammonia equivalent in the stereo selective ring-opening of chiral 2,3-epoxy alcohols (from Sharpless asymmetric epoxidation). The key step in the sequence involves intramolecular cyclization of the carbamate intermediate. It is also used in a multicomponent, stereospecific synthesis of 1,3-oxazinane-2,4-diones employing an Al-salph complex.

Solubility
It hydrolyzes with water.

Notes
Moisture& heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from alcohols, amines.
Specifications

Specifications

Benzyl isocyanate
1.078
43°C (109°F)
C8H7NO
C6H5CH2NCO
25 g
636508
benzyl isocyanate, isocyanatomethyl benzene, benzylisocyanate, isocyanic acid benzyl ester, benzene, isocyanatomethyl, 1-isocyanatomethyl benzene, phenylmethyl isocyanate, unii-8i2li357gq, phenylmethanisocyanate
YDNLNVZZTACNJX-UHFFFAOYSA-N
isocyanatomethylbenzene
76639
98%
3173-56-6
53°C to 54°C (0.8 mmHg)
Pungent
1.525
MFCD00009701
UN3080
Moisture sensitive
It hydrolyzes with water.
O=C=NCC1=CC=CC=C1
133.15
133.15

Chemical Identifiers

3173-56-6
133.15
YDNLNVZZTACNJX-UHFFFAOYSA-N
76639
O=C=NCC1=CC=CC=C1
C8H7NO
MFCD00009701
benzyl isocyanate, isocyanatomethyl benzene, benzylisocyanate, isocyanic acid benzyl ester, benzene, isocyanatomethyl, 1-isocyanatomethyl benzene, phenylmethyl isocyanate, unii-8i2li357gq, phenylmethanisocyanate
isocyanatomethylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H334-H226-H315-H319-H335
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P285-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P363-P370+P378q-P501c

H226-H301+H311+H331-H315-H317-H319-H334-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.

EINECSNumber : 221-640-9

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only