missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl methyl ether, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2349009

 View more versions of this product

Catalog No. AAB2349009


Explore available promotions

Description

Description

Benzyl methyl ether is used to produce benzaldehyde. It is used as perfuming agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl methyl ether is used to produce benzaldehyde. It is used as perfuming agents.

Solubility
Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

Benzyl methyl ether
-53°C
173°C to 175°C
C8H10O
MFCD00025901
UN3271
benzyl methyl ether, methoxymethyl benzene, methyl benzyl ether, benzene, methoxymethyl, ether, benzyl methyl, benzylmethylether, unii-f22rls78bd, .alpha.-methoxytoluene, f22rls78bd, methoxyphenylmethane
GQKZBCPTCWJTAS-UHFFFAOYSA-N
methoxymethylbenzene
10869
99%
538-86-3
0.94
56°C (133°F)
1.5015
10 g
14,1140
Insoluble in water.
COCC1=CC=CC=C1
122.167
122.17

Chemical Identifiers

538-86-3
122.167
GQKZBCPTCWJTAS-UHFFFAOYSA-N
10869
COCC1=CC=CC=C1
C8H10O
MFCD00025901
benzyl methyl ether, methoxymethyl benzene, methyl benzyl ether, benzene, methoxymethyl, ether, benzyl methyl, benzylmethylether, unii-f22rls78bd, .alpha.-methoxytoluene, f22rls78bd, methoxyphenylmethane
methoxymethylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c

H226-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 208-705-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only