missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzyl Methyl Sulfide 98.0+%, TCI America™
$108.62 - $331.79
Chemical Identifiers
CAS | 766-92-7 |
---|---|
Molecular Formula | C8H10S |
Molecular Weight (g/mol) | 138.228 |
MDL Number | MFCD00008563 |
InChI Key | OFQPKKGMNWASPN-UHFFFAOYSA-N |
Synonym | benzyl methyl sulfide, methyl benzyl sulfide, benzyl methyl sulphide, sulfide, benzyl methyl, 1-phenyl-2-thiapropane, benzene, methylthio methyl, alpha-methylthio toluene, methylthio methyl benzene, benzylmethylsulfide, unii-y3900rbk51 |
PubChem CID | 13016 |
IUPAC Name | methylsulfanylmethylbenzene |
SMILES | CSCC1=CC=CC=C1 |
Chemical Identifiers
766-92-7 | |
138.228 | |
OFQPKKGMNWASPN-UHFFFAOYSA-N | |
13016 | |
CSCC1=CC=CC=C1 |
C8H10S | |
MFCD00008563 | |
benzyl methyl sulfide, methyl benzyl sulfide, benzyl methyl sulphide, sulfide, benzyl methyl, 1-phenyl-2-thiapropane, benzene, methylthio methyl, alpha-methylthio toluene, methylthio methyl benzene, benzylmethylsulfide, unii-y3900rbk51 | |
methylsulfanylmethylbenzene |
Specifications
766-92-7 | |
200°C | |
MFCD00008563 | |
benzyl methyl sulfide, methyl benzyl sulfide, benzyl methyl sulphide, sulfide, benzyl methyl, 1-phenyl-2-thiapropane, benzene, methylthio methyl, alpha-methylthio toluene, methylthio methyl benzene, benzylmethylsulfide, unii-y3900rbk51 | |
CSCC1=CC=CC=C1 | |
138.228 | |
138.23 | |
Liquid |
Yellow | |
C8H10S | |
5 mL | |
OFQPKKGMNWASPN-UHFFFAOYSA-N | |
methylsulfanylmethylbenzene | |
13016 | |
≥98.0% (GC) | |
Benzyl Methyl Sulfide |
Safety and Handling
TSCA : Yes