missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl phenyl ether, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 310260250

 View more versions of this product

Catalog No. AC310260250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzyl phenyl ether
97.5
37°C to 41°C
286°C to 287°C
Authentic
Glass bottle
C6H5CH2OC6H5
25 g
Solubility in water: insoluble
C1=CC=C(C=C1)COC2=CC=CC=C2
184.24
184.24
Chunks
946-80-5
100.0
Brown-Orange
113°C
97.5% min. (GC)
C13H12O
MFCD00020660
benzyl phenyl ether, benzyloxy benzene, phenyl benzyl ether, benzene, phenoxymethyl, ether, benzyl phenyl, benzylphenylether, benzyloxy-benzene, benzyloxybenzene, unii-bue863n0l8, .alpha.-phenylanisole
BOTNYLSAWDQNEX-UHFFFAOYSA-N
phenoxymethylbenzene
70352
98%

Chemical Identifiers

946-80-5
184.24
BOTNYLSAWDQNEX-UHFFFAOYSA-N
70352
C1=CC=C(C=C1)COC2=CC=CC=C2
C13H12O
MFCD00020660
benzyl phenyl ether, benzyloxy benzene, phenyl benzyl ether, benzene, phenoxymethyl, ether, benzyl phenyl, benzylphenylether, benzyloxy-benzene, benzyloxybenzene, unii-bue863n0l8, .alpha.-phenylanisole
phenoxymethylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only