missing translation for 'onlineSavingsMsg'
Learn More

Benzyl propargyl ether, 97%

CAS: 4039-82-1 | C10H10O | 146.189 g/mol

$76.44 - $285.99

Chemical Identifiers

CAS 4039-82-1
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.189
MDL Number MFCD00483987
InChI Key PAQVEXAFKDWGOT-UHFFFAOYSA-N
Synonym benzyl propargyl ether, benzylpropargyl ether, 1-benzyloxy-2-propyne, prop-2-yn-1-yloxy methyl benzene, benzene, 2-propynyloxy methyl, prop-2-ynyloxy methyl benzene, propargylbenzyl ether, benzyloxy prop-2-yne
PubChem CID 6917484
IUPAC Name prop-2-ynoxymethylbenzene
SMILES C#CCOCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5348106
View Documents
Thermo Scientific Chemicals
H5348106
5 g
Each for $76.44
Only null left
 
AAH5348114
View Documents
Thermo Scientific Chemicals
H5348114
25 g
Each for $285.99
Only null left
 
Description

Description

Benzyl propargyl ether is used in the synthesis of substituted carbocyclic aromatic compounds and pharmaceutical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl propargyl ether is used in the synthesis of substituted carbocyclic aromatic compounds and pharmaceutical compounds.

Solubility
Soluble in chloroform.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

4039-82-1
146.189
PAQVEXAFKDWGOT-UHFFFAOYSA-N
6917484
C#CCOCC1=CC=CC=C1
C10H10O
MFCD00483987
benzyl propargyl ether, benzylpropargyl ether, 1-benzyloxy-2-propyne, prop-2-yn-1-yloxy methyl benzene, benzene, 2-propynyloxy methyl, prop-2-ynyloxy methyl benzene, propargylbenzyl ether, benzyloxy prop-2-yne
prop-2-ynoxymethylbenzene

Specifications

4039-82-1
101°C to 101°C (18 mmHg)
C6H4CH2OCH2C≡CH
MFCD00483987
benzyl propargyl ether, benzylpropargyl ether, 1-benzyloxy-2-propyne, prop-2-yn-1-yloxy methyl benzene, benzene, 2-propynyloxy methyl, prop-2-ynyloxy methyl benzene, propargylbenzyl ether, benzyloxy prop-2-yne
PAQVEXAFKDWGOT-UHFFFAOYSA-N
prop-2-ynoxymethylbenzene
6917484
97%
1.011
C10H10O
1.52
5 g
Soluble in chloroform.
C#CCOCC1=CC=CC=C1
146.189
146.19
Benzyl propargyl ether
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only