missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (triphenylphosphoranylidene)acetate, 97%

CAS: 15097-38-8 | C27H23O2P | 410.45 g/mol

$2599.37 - $2599.37

Chemical Identifiers

CAS 15097-38-8
Molecular Formula C27H23O2P
Molecular Weight (g/mol) 410.45
MDL Number MFCD00191787
InChI Key INKMLGJBBDRIQR-UHFFFAOYSA-N
Synonym benzyl 2-triphenylphosphoranylidene acetate, benzyl triphenylphosphoranylidene acetate, benzyl triphenylphosphoranylidene-acetate, benzyl 2-triphenyl-??-phosphanylidene acetate, acetic acid, triphenylphosphoranylidene-, phenylmethyl ester, phenylmethyl 2-triphenylylidene acetate, benzyl 2-triphenyl, acmc-20ah9g, ksc526q3l
PubChem CID 3862746
IUPAC Name benzyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
SMILES O=C(OCC1=CC=CC=C1)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC377221000
SDS View Documents
Thermo Scientific Chemicals
377221000
100 g Glass bottle
Each for $2,599.37
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

15097-38-8
410.45
INKMLGJBBDRIQR-UHFFFAOYSA-N
3862746
O=C(OCC1=CC=CC=C1)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C27H23O2P
MFCD00191787
benzyl 2-triphenylphosphoranylidene acetate, benzyl triphenylphosphoranylidene acetate, benzyl triphenylphosphoranylidene-acetate, benzyl 2-triphenyl-??-phosphanylidene acetate, acetic acid, triphenylphosphoranylidene-, phenylmethyl ester, phenylmethyl 2-triphenylylidene acetate, benzyl 2-triphenyl, acmc-20ah9g, ksc526q3l
benzyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate

Specifications

15097-38-8
100.0
White
96% min. (HPLC)
C27H23O2P
MFCD00191787
benzyl 2-triphenylphosphoranylidene acetate, benzyl triphenylphosphoranylidene acetate, benzyl triphenylphosphoranylidene-acetate, benzyl 2-triphenyl-??-phosphanylidene acetate, acetic acid, triphenylphosphoranylidene-, phenylmethyl ester, phenylmethyl 2-triphenylylidene acetate, benzyl 2-triphenyl, acmc-20ah9g, ksc526q3l
O=C(OCC1=CC=CC=C1)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
410.45
410.45
Powder
96.0
120.0°C to 127.0°C
Authentic
Glass bottle
(C6H5)3P=CHCOOCH2C6H5
100 g
INKMLGJBBDRIQR-UHFFFAOYSA-N
benzyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
3862746
97%
Benzyl (triphenylphosphoranylidene)acetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only