missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzylamine, 99%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 105850025

 View more versions of this product

Catalog No. AC105850025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzylamine
100-46-9
100.0
Colorless to Light Yellow
182.0°C to 185.0°C
Authentic
Glass bottle
1.5424 to 1.5444
MFCD00008106
12,1013
0.98
benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine
WGQKYBSKWIADBV-UHFFFAOYSA-N
phenylmethanamine
7504
CHEBI:40538
99%
Liquid
99%
98.5
-30.0°C
0.9800g/mL
72°C
98.5% min. (GC)
C7H9N
C6H5CH2NH2
2.5 L
01,51; 10,26; 13,79; 16,22
15,1128
Solubility in water: soluble in water. Other solubilities: >50g/100mL methanol,>50g/100mL ethanol,>50g/100mL 2-propanol,miscible with alcohol,ether,acetone,benzene
C1=CC=C(C=C1)CN
107.15
1.82 mPa.s (20°C)
107.15
Pure

Chemical Identifiers

100-46-9
107.15
WGQKYBSKWIADBV-UHFFFAOYSA-N
7504
phenylmethanamine
C7H9N
MFCD00008106
benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine
CHEBI:40538
C1=CC=C(C=C1)CN
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 202-854-1

RTECSNumber : DP1488500

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only