missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzylchlorodimethylsilane 98.0+%, TCI America™
Supplier: TCI America B23345G
Specifications
| Benzylchlorodimethylsilane | |
| Yellow | |
| C9H13ClSi | |
| 5 g | |
| ABHNFDUSOVXXOA-UHFFFAOYSA-N | |
| benzyl-chloro-dimethylsilane | |
| 15780 | |
| ≥98.0% (GC) |
| 1833-31-4 | |
| 211°C | |
| MFCD00018089 | |
| 2987 | |
| C[Si](C)(CC1=CC=CC=C1)Cl | |
| 184.738 | |
| 184.74 | |
| Liquid |
Chemical Identifiers
| 1833-31-4 | |
| 184.738 | |
| ABHNFDUSOVXXOA-UHFFFAOYSA-N | |
| benzyl-chloro-dimethylsilane |
| C9H13ClSi | |
| MFCD00018089 | |
| 15780 | |
| C[Si](C)(CC1=CC=CC=C1)Cl |
Safety and Handling
RTECSNumber : VV1810000
TSCA : Yes