missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzylidene-2-naphthylamine 98.0+%, TCI America™
Supplier: TCI America B026125G
Specifications
| Benzylidene-2-naphthylamine | |
| 101°C | |
| MFCD00059551 | |
| CKIGNOCMDJFFES-UHFFFAOYSA-N | |
| N-naphthalen-2-yl-1-phenylmethanimine | |
| 300779 | |
| ≥98.0% (N) |
| 891-32-7 | |
| C17H13N | |
| 25 g | |
| C1=CC=C(C=C1)C=NC2=CC3=CC=CC=C3C=C2 | |
| 231.298 | |
| 231.30 | |
| Crystalline Powder |
Chemical Identifiers
| 891-32-7 | |
| 231.298 | |
| CKIGNOCMDJFFES-UHFFFAOYSA-N | |
| N-naphthalen-2-yl-1-phenylmethanimine |
| C17H13N | |
| MFCD00059551 | |
| 300779 | |
| C1=CC=C(C=C1)C=NC2=CC3=CC=CC=C3C=C2 |
Safety and Handling
TSCA : No