Learn More
Benzyltriethylammonium chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 164021000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Benzyltriethylammonium chloride | |
56-37-1 | |
100.0 | |
White to Beige | |
Authentic | |
Glass bottle | |
C6H5CH2N(C2H5)3Cl | |
100 g | |
Solubility in water: 700g/L water (20°C). Other solubilities: soluble in methanol,ethanol,isopropanol,dmf,soluble in acetone,ch2cl2 | |
[Cl-].CC[N+](CC)(CC)CC1=CC=CC=C1 | |
227.78 | |
227.78 | |
Crystalline Powder |
98% | |
97.5 | |
185.0°C | |
>275°C | |
97.5 to 102.5% (Argentometry) | |
C13H22ClN | |
MFCD00011824 | |
benzyltriethylammonium chloride, tebac, benzyl triethylammonium chloride, benzyl triethyl ammonium chloride, benzyltriethylammoniumchloride, triethylbenzylammonium chloride, bteac, teba, benzyltriethylazanium chloride, n-benzyl-n,n-diethylethanaminium chloride | |
HTZCNXWZYVXIMZ-UHFFFAOYSA-M | |
benzyl(triethyl)azanium;chloride | |
66133 | |
98% |
Chemical Identifiers
56-37-1 | |
227.78 | |
HTZCNXWZYVXIMZ-UHFFFAOYSA-M | |
66133 | |
[Cl-].CC[N+](CC)(CC)CC1=CC=CC=C1 |
C13H22ClN | |
MFCD00011824 | |
benzyltriethylammonium chloride, tebac, benzyl triethylammonium chloride, benzyl triethyl ammonium chloride, benzyltriethylammoniumchloride, triethylbenzylammonium chloride, bteac, teba, benzyltriethylazanium chloride, n-benzyl-n,n-diethylethanaminium chloride | |
benzyl(triethyl)azanium;chloride |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-270-1
RTECSNumber : BO8275000
TSCA : TSCA
RUO – Research Use Only