missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltriethylammonium chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 164025000

 View more versions of this product

Catalog No. AC164025000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Benzyltriethylammonium chloride
56-37-1
100.0
White to Beige
Authentic
Glass bottle
C6H5CH2N(C2H5)3Cl
500 g
Solubility in water: 700g/L water (20°C). Other solubilities: soluble in methanol,ethanol,isopropanol,dmf,soluble in acetone,ch2cl2
[Cl-].CC[N+](CC)(CC)CC1=CC=CC=C1
227.78
227.78
Crystalline Powder
98%
97.5
185.0°C
>275°C
97.5 to 102.5% (Argentometry)
C13H22ClN
MFCD00011824
benzyltriethylammonium chloride, tebac, benzyl triethylammonium chloride, benzyl triethyl ammonium chloride, benzyltriethylammoniumchloride, triethylbenzylammonium chloride, bteac, teba, benzyltriethylazanium chloride, n-benzyl-n,n-diethylethanaminium chloride
HTZCNXWZYVXIMZ-UHFFFAOYSA-M
benzyl(triethyl)azanium;chloride
66133
98%

Chemical Identifiers

56-37-1
227.78
HTZCNXWZYVXIMZ-UHFFFAOYSA-M
66133
[Cl-].CC[N+](CC)(CC)CC1=CC=CC=C1
C13H22ClN
MFCD00011824
benzyltriethylammonium chloride, tebac, benzyl triethylammonium chloride, benzyl triethyl ammonium chloride, benzyltriethylammoniumchloride, triethylbenzylammonium chloride, bteac, teba, benzyltriethylazanium chloride, n-benzyl-n,n-diethylethanaminium chloride
benzyl(triethyl)azanium;chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-270-1

RTECSNumber : BO8275000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only