Learn More
Benzyltrimethylammonium bromide, 98+%, Thermo Scientific Chemicals
$58.95 - $58.95
Chemical Identifiers
CAS | 5350-41-4 |
---|---|
Molecular Formula | C10H16BrN |
Molecular Weight (g/mol) | 230.15 |
MDL Number | MFCD00011780 |
InChI Key | UUZYBYIOAZTMGC-UHFFFAOYSA-M |
Synonym | benzyltrimethylammonium bromide, n,n,n-trimethyl-1-phenylmethanaminium bromide, trimethylbenzylammonium bromide, n-benzyl-n,n,n-trimethylammonium bromide, wv 562 german, benzenemethanaminium, n,n,n-trimethyl-, bromide, nsc 24, ammonium, benzyltrimethyl-, bromide, benzyltrimethylammonium bromide btm |
PubChem CID | 21449 |
IUPAC Name | benzyl(trimethyl)azanium;bromide |
SMILES | C[N+](C)(C)CC1=CC=CC=C1.[Br-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC364860250
|
Thermo Scientific Chemicals
364860250 |
25 g | Glass bottle |
Each for $58.95
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
5350-41-4 | |
230.15 | |
UUZYBYIOAZTMGC-UHFFFAOYSA-M | |
21449 | |
C[N+](C)(C)CC1=CC=CC=C1.[Br-] |
C10H16BrN | |
MFCD00011780 | |
benzyltrimethylammonium bromide, n,n,n-trimethyl-1-phenylmethanaminium bromide, trimethylbenzylammonium bromide, n-benzyl-n,n,n-trimethylammonium bromide, wv 562 german, benzenemethanaminium, n,n,n-trimethyl-, bromide, nsc 24, ammonium, benzyltrimethyl-, bromide, benzyltrimethylammonium bromide btm | |
benzyl(trimethyl)azanium;bromide |
Specifications
5350-41-4 | |
100.0 | |
White | |
Authentic | |
Glass bottle | |
MFCD00011780 | |
benzyltrimethylammonium bromide, n,n,n-trimethyl-1-phenylmethanaminium bromide, trimethylbenzylammonium bromide, n-benzyl-n,n,n-trimethylammonium bromide, wv 562 german, benzenemethanaminium, n,n,n-trimethyl-, bromide, nsc 24, ammonium, benzyltrimethyl-, bromide, benzyltrimethylammonium bromide btm | |
UUZYBYIOAZTMGC-UHFFFAOYSA-M | |
benzyl(trimethyl)azanium;bromide | |
21449 | |
98+% | |
Benzyltrimethylammonium bromide, 99% |
98.5 | |
228°C to 234°C | |
1% max. | |
98% min. (Argentometry) | |
C10H16BrN | |
25 g | |
Solubility in water: soluble | |
C[N+](C)(C)CC1=CC=CC=C1.[Br-] | |
230.15 | |
230.15 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 226-320-2
RUO – Research Use Only