Learn More
Benzyltrimethylammonium bromide, 98+%, Thermo Scientific Chemicals
$64.66 - $175.68
Chemical Identifiers
CAS | 5350-41-4 |
---|---|
Molecular Formula | C10H16BrN |
Molecular Weight (g/mol) | 230.149 |
MDL Number | MFCD00011780 |
InChI Key | UUZYBYIOAZTMGC-UHFFFAOYSA-M |
Synonym | benzyltrimethylammonium bromide, n,n,n-trimethyl-1-phenylmethanaminium bromide, trimethylbenzylammonium bromide, n-benzyl-n,n,n-trimethylammonium bromide, wv 562 german, benzenemethanaminium, n,n,n-trimethyl-, bromide, nsc 24, ammonium, benzyltrimethyl-, bromide, benzyltrimethylammonium bromide btm |
PubChem CID | 21449 |
IUPAC Name | benzyl(trimethyl)azanium;bromide |
SMILES | C[N+](C)(C)CC1=CC=CC=C1.[Br-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1684014
|
Thermo Scientific Chemicals
A1684014 |
25 g |
Each for $64.66
|
|
|||||
AAA1684022
|
Thermo Scientific Chemicals
A1684022 |
100 g |
Each for $175.68
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5350-41-4 | |
230.149 | |
UUZYBYIOAZTMGC-UHFFFAOYSA-M | |
21449 | |
C[N+](C)(C)CC1=CC=CC=C1.[Br-] |
C10H16BrN | |
MFCD00011780 | |
benzyltrimethylammonium bromide, n,n,n-trimethyl-1-phenylmethanaminium bromide, trimethylbenzylammonium bromide, n-benzyl-n,n,n-trimethylammonium bromide, wv 562 german, benzenemethanaminium, n,n,n-trimethyl-, bromide, nsc 24, ammonium, benzyltrimethyl-, bromide, benzyltrimethylammonium bromide btm | |
benzyl(trimethyl)azanium;bromide |
Specifications
5350-41-4 | |
C10H16BrN | |
25 g | |
Hygroscopic | |
UUZYBYIOAZTMGC-UHFFFAOYSA-M | |
benzyl(trimethyl)azanium;bromide | |
21449 | |
≥98% |
228°C to 232°C | |
MFCD00011780 | |
3917251 | |
benzyltrimethylammonium bromide, n,n,n-trimethyl-1-phenylmethanaminium bromide, trimethylbenzylammonium bromide, n-benzyl-n,n,n-trimethylammonium bromide, wv 562 german, benzenemethanaminium, n,n,n-trimethyl-, bromide, nsc 24, ammonium, benzyltrimethyl-, bromide, benzyltrimethylammonium bromide btm | |
C[N+](C)(C)CC1=CC=CC=C1.[Br-] | |
230.149 | |
230.15 | |
Benzyltrimethylammonium bromide |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 226-320-2
RTECSNumber : BO8392000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only