missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltrimethylammonium chloride, 50 wt.% solution in water, Thermo Scientific Chemicals

$49.94 - $205.18

Chemical Identifiers

CAS 56-93-9
Molecular Formula C10H16ClN
Molecular Weight (g/mol) 185.70
MDL Number MFCD00011782
InChI Key KXHPPCXNWTUNSB-UHFFFAOYSA-M
Synonym benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride
PubChem CID 5963
IUPAC Name benzyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC209381000
SDS View Documents
Thermo Scientific Chemicals
209381000
100 g Glass bottle
Each for $49.94
 
AC209380010
SDS View Documents
Thermo Scientific Chemicals
209380010
1 kg Glass bottle
Each for $205.18
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Chemical Identifiers

56-93-9
185.70
KXHPPCXNWTUNSB-UHFFFAOYSA-M
5963
[Cl-].C[N+](C)(C)CC1=CC=CC=C1
C10H16ClN
MFCD00011782
benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride
benzyl(trimethyl)azanium;chloride

Specifications

Colorless to Yellow
49.0,49.0
C10H16ClN
MFCD00011782
01,53
benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride
[Cl-].C[N+](C)(C)CC1=CC=CC=C1
185.70
185.7
Glass bottle
1.044
Liquid
56-93-9,7732-18-5
51.0,51.0
C6H5CH2N(CH3)3Cl
12, 1020
1.0440g/mL
KXHPPCXNWTUNSB-UHFFFAOYSA-M
benzyl(trimethyl)azanium;chloride
5963
≥49%
1.445
100 g
Benzyltrimethylammonium chloride, 50 wt. % Solution in Water
Videos
Safety and Handling

Safety and Handling

GHS P Statement Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Suspected of causing genetic defects. Harmful to aquatic life with long lasting effects.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF exposed: Call a POISON CENTRE or doctor/physician. Avoid release to the environment.

Danger

EINECSNumber : 200-300-3

RTECSNumber : BO8400000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only