missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 211860500

 View more versions of this product

Catalog No. AC211860500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Benzyltrimethylammonium chloride
56-93-9
100.0
White to Ivory
98+%
C10H16ClN
MFCD00011782
12, 1020
benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride
KXHPPCXNWTUNSB-UHFFFAOYSA-M
benzyl(trimethyl)azanium;chloride
5963
185.7
Crystalline Powder and/or Chunks
98+%
98.0
239.0°C
Authentic
Glass bottle
C6H5CH2N(CH3)3Cl
50 g
01,53
Solubility in water: 800g/L. Other solubilities: soluble in ethanol and butanol,slightly soluble in butyl phthalate and,tributyl phosphate
[Cl-].C[N+](C)(C)CC1=CC=CC=C1
185.70
10 mPa.s (20°C)
98+%

Chemical Identifiers

56-93-9
185.70
KXHPPCXNWTUNSB-UHFFFAOYSA-M
5963
[Cl-].C[N+](C)(C)CC1=CC=CC=C1
C10H16ClN
MFCD00011782
benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride
benzyl(trimethyl)azanium;chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Suspected of causing genetic defects.
Harmful to aquatic life with long lasting effects.
Toxic by eye contact.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse

GHS Signal Word: Danger

EINECSNumber : 200-300-3

RTECSNumber : BO8400000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only