missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltrimethylammonium dichloroiodate, 95%, Thermo Scientific Chemicals

$84.18 - $270.27

Chemical Identifiers

CAS 114971-52-7
Molecular Formula C10H16Cl2IN
Molecular Weight (g/mol) 348.049
MDL Number MFCD00075259
InChI Key PPDJNZTUDFPAHX-UHFFFAOYSA-N
Synonym benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, btma-icl2, benzyl trimethylammonium dichloroiodate, benzyltrimethylazanium; dichloroiodanuide, pubchem21920, benzyltrimethylammoniumdichloroiodate
PubChem CID 2724680
SMILES C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2289206
View Documents
Thermo Scientific Chemicals
B2289206
5 g
Each for $84.18
 
AAB2289214
View Documents
Thermo Scientific Chemicals
B2289214
25 g
Each for $270.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

114971-52-7
348.049
PPDJNZTUDFPAHX-UHFFFAOYSA-N
2724680
C10H16Cl2IN
MFCD00075259
benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, btma-icl2, benzyl trimethylammonium dichloroiodate, benzyltrimethylazanium; dichloroiodanuide, pubchem21920, benzyltrimethylammoniumdichloroiodate
C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl

Specifications

114971-52-7
C10H16Cl2IN
MFCD00075259
4611949
benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, btma-icl2, benzyl trimethylammonium dichloroiodate, benzyltrimethylazanium; dichloroiodanuide, pubchem21920, benzyltrimethylammoniumdichloroiodate
C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl
2724680
95%
∼125°C
C6H5CH2(CH3)3NICl2
5 g
Hygroscopic; Light sensitive
PPDJNZTUDFPAHX-UHFFFAOYSA-N
348.049
348.06
Benzyltrimethylammonium dichloroiodate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only