missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltrimethylammonium Dichloroiodate 97.0+%, TCI America™

$37.52 - $106.65

Chemical Identifiers

CAS 114971-52-7
Molecular Formula C10H16Cl2IN
Molecular Weight (g/mol) 348.049
MDL Number MFCD00075259
InChI Key PPDJNZTUDFPAHX-UHFFFAOYSA-N
Synonym benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, btma-icl2, benzyl trimethylammonium dichloroiodate, benzyltrimethylazanium; dichloroiodanuide, pubchem21920, benzyltrimethylammoniumdichloroiodate
PubChem CID 2724680
SMILES C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B16045G
View Documents
TCI America
B16045G
5 g
Each for $37.52
 
B160425G
View Documents
TCI America
B160425G
25 g
Each for $106.65
 
Description

Description

Specifications

Chemical Identifiers

114971-52-7
348.049
PPDJNZTUDFPAHX-UHFFFAOYSA-N
2724680
C10H16Cl2IN
MFCD00075259
benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, btma-icl2, benzyl trimethylammonium dichloroiodate, benzyltrimethylazanium; dichloroiodanuide, pubchem21920, benzyltrimethylammoniumdichloroiodate
C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl

Specifications

114971-52-7
C10H16Cl2IN
5 g
PPDJNZTUDFPAHX-UHFFFAOYSA-N
348.049
348.05
Crystalline Powder
125°C
MFCD00075259
benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, btma-icl2, benzyl trimethylammonium dichloroiodate, benzyltrimethylazanium; dichloroiodanuide, pubchem21920, benzyltrimethylammoniumdichloroiodate
C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl
2724680
≥97.0% (T)
Benzyltrimethylammonium Dichloroiodate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2694

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'