missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltrimethylammonium hydroxide, 40 wt% solution in methanol, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 106200051

Catalog No. AC106200051

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Phase transfer catalyst
Specifications

Specifications

Benzyltrimethylammonium hydroxide
100-85-6,67-56-1
41.0,61.0
C6H5CH2N(CH3)3OH
01,1252; 05,29; 08,36; 12,48; 05,30
benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide
C[N+](C)(C)CC1=CC=CC=C1.[OH-]
167.252
167.25
Plastic Drum
5L
40 wt. %solution in Methanol
39.0,59.0
C10H17NO
MFCD00008281
0.9250g/mL
NDKBVBUGCNGSJJ-UHFFFAOYSA-M
benzyl(trimethyl)azanium;hydroxide
66854
≥40%
0.925
15°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 202-895-5

RTECSNumber : BO8575000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only