missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyltrimethylammonium hydroxide, 40 wt% solution in methanol, Thermo Scientific Chemicals

$144.34 - $369.47

Chemical Identifiers

CAS 100-85-6
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD00008281
InChI Key NDKBVBUGCNGSJJ-UHFFFAOYSA-M
Synonym benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide
PubChem CID 66854
IUPAC Name benzyl(trimethyl)azanium;hydroxide
SMILES C[N+](C)(C)CC1=CC=CC=C1.[OH-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC106202500
SDS View Documents
Thermo Scientific Chemicals
106202500
250 mL Glass bottle
Each for $144.34
 
AC106200010
SDS View Documents
Thermo Scientific Chemicals
106200010
1 L Glass bottle
Each for $369.47
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Chemical Identifiers

100-85-6
167.252
NDKBVBUGCNGSJJ-UHFFFAOYSA-M
66854
C[N+](C)(C)CC1=CC=CC=C1.[OH-]
C10H17NO
MFCD00008281
benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide
benzyl(trimethyl)azanium;hydroxide

Specifications

Yellow
39.0,59.0
C10H17NO
MFCD00008281
0.9250g/mL
Solubility in water: miscible.
C[N+](C)(C)CC1=CC=CC=C1.[OH-]
167.252
167.25
Glass bottle
250 mL
Solution
100-85-6,67-56-1
41.0,61.0
C6H5CH2N(CH3)3OH
01,1252; 05,29; 08,36; 12,48; 05,30
benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide
NDKBVBUGCNGSJJ-UHFFFAOYSA-M
benzyl(trimethyl)azanium;hydroxide
66854
≥40%
0.925
15°C
Benzyltrimethylammonium hydroxide, 40 wt. %solution in Methanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 202-895-5

RTECSNumber : BO8575000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only