missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Beryllium foil, 0.25mm (0.01in) thick, 99.8% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 041642FF

 View more versions of this product

Catalog No. AA41642FF


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Beryllium foil
7440-41-7
Be
25 x 25 mm
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
beryllium
5460467
99.8%
≈0.29g/25x25mm
0.25mm (0.01 in.) thick, Vacuum tight
(metals basis)
MFCD00134032
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
[Be]
9.01
CHEBI:33783
Odorless
Foil

Chemical Identifiers

7440-41-7
9.01
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
5460467
beryllium
Be
MFCD00134032
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
CHEBI:33783
[Be]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P362-P501c

H301-H315-H317-H319-H330-H335-H350i-H372

EINECSNumber : 231-150-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only