missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Beryllium lump, irregular, 3-38mm (0.1-1.5in), 99.5% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01285318
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gray | |
7440-41-7 | |
99.5% (Metals basis) | |
MFCD00134032 | |
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds | |
[Be] | |
9.01 | |
CHEBI:33783 | |
Lump |
Beryllium | |
Gray irregular lump | |
Be | |
50 g | |
ATBAMAFKBVZNFJ-UHFFFAOYSA-N | |
beryllium | |
5460467 | |
Odorless |
Chemical Identifiers
7440-41-7 | |
9.01 | |
ATBAMAFKBVZNFJ-UHFFFAOYSA-N | |
5460467 | |
beryllium |
Be | |
MFCD00134032 | |
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds | |
CHEBI:33783 | |
[Be] |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P362-P501c
H301-H315-H317-H319-H330-H335-H350i-H372
EINECSNumber : 231-150-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only