Learn More
Beryllium wire, 0.25mm (0.01 in.) dia., annealed, 99.7% (metals basis), Thermo Scientific Chemicals
$1102.04 - $2074.37
Chemical Identifiers
CAS | 7440-41-7 |
---|---|
Molecular Formula | Be |
Molecular Weight (g/mol) | 9.01 |
MDL Number | MFCD00134032 |
InChI Key | ATBAMAFKBVZNFJ-UHFFFAOYSA-N |
Synonym | glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds |
PubChem CID | 5460467 |
ChEBI | CHEBI:33783 |
IUPAC Name | beryllium |
SMILES | [Be] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-41-7 | |
9.01 | |
ATBAMAFKBVZNFJ-UHFFFAOYSA-N | |
5460467 | |
beryllium |
Be | |
MFCD00134032 | |
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds | |
CHEBI:33783 | |
[Be] |
Specifications
7440-41-7 | |
MFCD00134032 | |
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds | |
[Be] | |
9.01 | |
CHEBI:33783 | |
Odorless | |
Wire | |
(metals basis) |
Be | |
25 cm | |
ATBAMAFKBVZNFJ-UHFFFAOYSA-N | |
beryllium | |
5460467 | |
99.7% | |
≈0.091g/m | |
Beryllium wire, 0.25mm (0.01 in.) dia., Annealed, Temper: soft |
Safety and Handling
GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P362-P501c
H301-H315-H317-H319-H330-H335-H350i-H372
EINECSNumber : 231-150-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only