CMD (Cotton Mouton Diagnostics) αBET Instrument
The CMD αBET system is a fully integrated testing system providing a more rapid and sustainable solution to endotoxin testing without compromising on sensitivity or performance.
Supplier:  CMD (Cotton Mouton Diagnostics) CMDA0007EN
Catalog No. 12-333-033
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Novel technology incorporating magneto-optical detection methods
  • Compact and portable
  • Highly flexible - allows for pharmacopoeial compliant assays or simple sample analysis
  • 4-channel simultaneous testing
  • Enhanced performance using a fraction of FDA-licensed turbidimetric LAL reagent
  • Quantitative results are provided in less than half the time of traditional assays
  • Minimal user input is needed beyond the introduction of the sample
  • On-board FDA Part 11 ready software package ensures data integrity
  • Highly sensitive - able to test down to 0.001EU/mL