missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Alaninamide Hydrochloride 98.0+%, TCI America™
$331.81 - $1258.24
Chemical Identifiers
| CAS | 64017-81-8 |
|---|---|
| Molecular Formula | C3H9ClN2O |
| Molecular Weight (g/mol) | 124.568 |
| MDL Number | MFCD00060747 |
| InChI Key | GAGJMOQGABUOBK-UHFFFAOYSA-N |
| Synonym | 3-aminopropanamide hydrochloride, 3-aminopropionamide hydrochloride, 3-aminopropanamide hcl, beta-alaninamide hydrochloride, h-beta-ala-nh2.hcl, h-beta-ala-nh2 hcl, 3-amino-propionamide hydrochloride, h-ala-nh2hcl, propanamide, 3-amino-, monohydrochloride, h-beta-ala-nh2hcl |
| PubChem CID | 22222288 |
| IUPAC Name | 3-aminopropanamide;hydrochloride |
| SMILES | C(CN)C(=O)N.Cl |
Chemical Identifiers
| 64017-81-8 | |
| 124.568 | |
| GAGJMOQGABUOBK-UHFFFAOYSA-N | |
| 22222288 | |
| C(CN)C(=O)N.Cl |
| C3H9ClN2O | |
| MFCD00060747 | |
| 3-aminopropanamide hydrochloride, 3-aminopropionamide hydrochloride, 3-aminopropanamide hcl, beta-alaninamide hydrochloride, h-beta-ala-nh2.hcl, h-beta-ala-nh2 hcl, 3-amino-propionamide hydrochloride, h-ala-nh2hcl, propanamide, 3-amino-, monohydrochloride, h-beta-ala-nh2hcl | |
| 3-aminopropanamide;hydrochloride |
Specifications
| 153°C | |
| White | |
| MFCD00060747 | |
| GAGJMOQGABUOBK-UHFFFAOYSA-N | |
| 3-aminopropanamide;hydrochloride | |
| 124.568 | |
| 124.57 | |
| Crystalline Powder |
| 64017-81-8 | |
| C3H9ClN2O | |
| 3-aminopropanamide hydrochloride, 3-aminopropionamide hydrochloride, 3-aminopropanamide hcl, beta-alaninamide hydrochloride, h-beta-ala-nh2.hcl, h-beta-ala-nh2 hcl, 3-amino-propionamide hydrochloride, h-ala-nh2hcl, propanamide, 3-amino-, monohydrochloride, h-beta-ala-nh2hcl | |
| C(CN)C(=O)N.Cl | |
| 1 g | |
| 22222288 | |
| ≥98.0% (T) | |
| beta-Alaninamide Hydrochloride |
Safety and Handling
TSCA : No