missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine methyl ester hydrochloride, 98%

CAS: 3196-73-4 | C4H10ClNO2 | 139.58 g/mol

$123.58 - $340.70

Chemical Identifiers

CAS 3196-73-4
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00039060
InChI Key XPGRZDJXVKFLHQ-UHFFFAOYSA-N
Synonym methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, a-ala-ome.hcl, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride
PubChem CID 2734767
IUPAC Name methyl 3-aminopropanoate;hydrochloride
SMILES Cl.COC(=O)CCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2748306
View Documents
Thermo Scientific Chemicals
H2748306
5 g
Each for $123.58
Only null left
 
AAH2748314
View Documents
Thermo Scientific Chemicals
H2748314
25 g
Each for $340.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3196-73-4
139.58
XPGRZDJXVKFLHQ-UHFFFAOYSA-N
2734767
Cl.COC(=O)CCN
C4H10ClNO2
MFCD00039060
methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, a-ala-ome.hcl, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride
methyl 3-aminopropanoate;hydrochloride

Specifications

102°C to 104°C
C4H10ClNO2
MFCD00039060
methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, a-ala-ome.hcl, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride
Cl.COC(=O)CCN
5 g
2734767
Hygroscopic
beta-Alanine methyl ester hydrochloride
3196-73-4
H2NCH2CH2CO2CH3·HCl
3556748
XPGRZDJXVKFLHQ-UHFFFAOYSA-N
methyl 3-aminopropanoate;hydrochloride
139.58
139.58
98%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only