missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Alanine Methyl Ester Hydrochloride 98.0+%, TCI America™
$83.89 - $288.94
Chemical Identifiers
| CAS | 3196-73-4 |
|---|---|
| Molecular Formula | C4H10ClNO2 |
| Molecular Weight (g/mol) | 139.58 |
| MDL Number | MFCD00039060 |
| InChI Key | XPGRZDJXVKFLHQ-UHFFFAOYSA-N |
| Synonym | methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, h-beta-ala-ome.hcl, h-beta-ala-ome.hcl, h-, a-ala-ome.hcl, b-alanine methyl ester hydrochloride, b-alanine methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride |
| PubChem CID | 2734767 |
| IUPAC Name | methyl 3-aminopropanoate hydrochloride |
| SMILES | Cl.COC(=O)CCN |
Chemical Identifiers
| 3196-73-4 | |
| 139.58 | |
| XPGRZDJXVKFLHQ-UHFFFAOYSA-N | |
| 2734767 | |
| Cl.COC(=O)CCN |
| C4H10ClNO2 | |
| MFCD00039060 | |
| methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, h-beta-ala-ome.hcl, h-beta-ala-ome.hcl, h-, a-ala-ome.hcl, b-alanine methyl ester hydrochloride, b-alanine methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride | |
| methyl 3-aminopropanoate hydrochloride |
Specifications
| 108°C | |
| White-Yellow | |
| MFCD00039060 | |
| XPGRZDJXVKFLHQ-UHFFFAOYSA-N | |
| methyl 3-aminopropanoate hydrochloride | |
| 139.58 | |
| 139.58 | |
| Crystalline Powder |
| 3196-73-4 | |
| C4H10ClNO2 | |
| methyl 3-aminopropionate hydrochloride, 3-aminopropionic acid methyl ester hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, b-alanine methyl ester hydrochloride, b-alanine methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride | |
| Cl.COC(=O)CCN | |
| 5 g | |
| 2734767 | |
| ≥98.0% (T) | |
| beta-Alanine Methyl Ester Hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator