missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals beta-Casomorphin, human, 95+%

An opioid peptide

$121.27 - $420.03

Chemical Identifiers

CAS 102029-74-3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62271LB0
View Documents
Thermo Scientific Chemicals
J62271LB0
5mg
Each for $121.27
 
AAJ62271MB
View Documents
Thermo Scientific Chemicals
J62271MB
25mg
Each for $420.03
 
Description

Description

An opioid peptide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

102029-74-3

Specifications

≥95%
White
Powder
beta-Casomorphin
102029-74-3
Store at -20°C
864.01
5mg
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only