missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
beta-Citronellol 95.0+%, TCI America™
Supplier: TCI America C0370500ML
Specifications
beta-Citronellol | |
Yellow | |
C10H20O | |
500 mL | |
QMVPMAAFGQKVCJ-UHFFFAOYSA-N | |
3,7-dimethyloct-6-en-1-ol | |
8842 | |
156.27 | |
Liquid |
106-22-9 | |
220°C | |
MFCD00002935 | |
citronellol, beta-citronellol, 3,7-dimethyl-6-octen-1-ol, dl-citronellol, cephrol, elenol, 6-octen-1-ol, 3,7-dimethyl, rodinol, 2,3-dihydrogeraniol, 2,6-dimethyl-2-octen-8-ol | |
CC(CCC=C(C)C)CCO | |
156.269 | |
CHEBI:50462 | |
≥95.0% (GC) |
Chemical Identifiers
106-22-9 | |
156.269 | |
QMVPMAAFGQKVCJ-UHFFFAOYSA-N | |
8842 | |
3,7-dimethyloct-6-en-1-ol |
C10H20O | |
MFCD00002935 | |
citronellol, beta-citronellol, 3,7-dimethyl-6-octen-1-ol, dl-citronellol, cephrol, elenol, 6-octen-1-ol, 3,7-dimethyl, rodinol, 2,3-dihydrogeraniol, 2,6-dimethyl-2-octen-8-ol | |
CHEBI:50462 | |
CC(CCC=C(C)C)CCO |
Safety and Handling
EINECSNumber : (2)-0258
RTECSNumber : RH3400000
TSCA : Yes