Learn More
beta-Cyclodextrin hydrate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1452906
Description
beta-Cyclodextrin hydrate forms clathrates. Produces a water-soluble complex with dansyl chloride for the fluorescent labeling of proteins. It is used in pharmaceutical research, as pharmaceutical intermediates and also as chemical reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsbeta-Cyclodextrin hydrate forms clathrates. Produces a water-soluble complex with dansyl chloride for the fluorescent labeling of proteins. It is used in pharmaceutical research, as pharmaceutical intermediates and also as chemical reagents.
Solubility
Slightly soluble in water (18.5 g/L at 25°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
beta-Cyclodextrin hydrate | |
288°C (decomposition) | |
MFCD00150811 | |
5915513 | |
beta-cyclodextrin hydrate, schardinger, a-dextrin hydrate, beta-cyclodextrin hydrate, puriss hplc | |
WHGYBXFWUBPSRW-UHFFFAOYSA-N | |
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O | |
1134.99 | |
1134.98 (Anhydrous) |
68168-23-0 | |
C42H70O35 | |
5 g | |
14,2718 | |
Slightly soluble in water (18.5g/L at 25°C). | |
+142° (c=1.5 in Water) | |
5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,â¶.2â¸,¹¹.2¹³,¹â¶.2¹â¸,²¹.2²³,²â¶.2²â¸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol | |
53486154 |
Chemical Identifiers
68168-23-0 | |
1134.99 | |
WHGYBXFWUBPSRW-UHFFFAOYSA-N | |
53486154 | |
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O |
C42H70O35 | |
MFCD00150811 | |
beta-cyclodextrin hydrate, schardinger, a-dextrin hydrate, beta-cyclodextrin hydrate, puriss hplc | |
5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,â¶.2â¸,¹¹.2¹³,¹â¶.2¹â¸,²¹.2²³,²â¶.2²â¸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol |
Safety and Handling
EINECSNumber : 231-493-2
RTECSNumber : GU2293000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only