Learn More
Thermo Scientific Chemicals beta-D-Gentiobiose, 98%
CAS: 554-91-6 | C12H22O11
Supplier: Thermo Scientific Chemicals J66750ME
Description
Gentiobiose was used in binding assays to determine if coelomic cytolytic factor 1 (CCF-1) binds to β-1,3-glucans to stimulate the pro-PO-activating system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGentiobiose was used in binding assays to determine if coelomic cytolytic factor 1 (CCF-1) binds to β-1,3-glucans to stimulate the pro-PO-activating system.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
| beta-D-Gentiobiose | |
| White | |
| MFCD00198056 | |
| 93354 | |
| Slightly soluble in water. | |
| 98% |
| 554-91-6 | |
| C12H22O11 | |
| 500 mg | |
| 6-O-(beta-D-Glucopyranosyl)-D-glucopyranose; Amygdalose | |
| 342.3 | |
| Powder |
Chemical Identifiers
| 554-91-6 | |
| C12H22O11 | |
| MFCD00198056 | |
| 6-O-(beta-D-Glucopyranosyl)-D-glucopyranose; Amygdalose |
Safety and Handling
EINECSNumber : 209-074-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only