missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

beta-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%, Thermo Scientific Chemicals

$76.52 - $790.74

Chemical Identifiers

CAS 13035-61-5
Molecular Formula C13H18O9
Molecular Weight (g/mol) 318.278
MDL Number MFCD00005358
InChI Key IHNHAHWGVLXCCI-FDYHWXHSSA-N
Synonym beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d
PubChem CID 83064
IUPAC Name [(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2442606
View Documents
Thermo Scientific Chemicals
B2442606
5 g
Each for $76.52
 
AAB2442614
View Documents
Thermo Scientific Chemicals
B2442614
25 g
Each for $252.01
 
AAB2442622
View Documents
Thermo Scientific Chemicals
B2442622
100 g
Each for $790.74
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13035-61-5
318.278
IHNHAHWGVLXCCI-FDYHWXHSSA-N
83064
CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C
C13H18O9
MFCD00005358
beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d
[(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate

Specifications

13035-61-5
C13H18O9
5 g
beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d
−13±0.5° (c=2.4 in Chloroform)
[(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
83064
≥98%
80°C to 83°C
MFCD00005358
94078
IHNHAHWGVLXCCI-FDYHWXHSSA-N
CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C
318.278
318.28
beta-D-Ribofuranose 1,2,3,5-tetraacetate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 235-898-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only