missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

beta-Glycerophosphate, 200mM soln., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J62121AE

 View more versions of this product

Catalog No. AAJ62121AE


Explore available promotions

Description

Description

β-Glycerolphosphate (Glycerol-2-Phosphate) is used in the development of hydrogels and scaffolds that have applications in tissue engineering and cell growth and differentiation. It has also been used as an additive in isolation mediums, similarly providing available phosphate to the isolate. It is a phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor. Accelerates calcification in vascular smooth muscle cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
β-Glycerolphosphate (Glycerol-2-Phosphate) is used in the development of hydrogels and scaffolds that have applications in tissue engineering and cell growth and differentiation. It has also been used as an additive in isolation mediums, similarly providing available phosphate to the isolate. It is a phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor. Accelerates calcification in vascular smooth muscle cells.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

beta-Glycerophosphate
Colorless
MFCD00002135
glycerol 2-phosphate disodium salt hydrate, disodium glycerol 2-phosphate hydrate, beta-glycerophosphoric acid disodium salt, c3h7o6p.2na.h2o, sodium glycerophosphate usan, b-glycerophosphate disodium salt hydrate, sodium 1,3-dihydroxypropan-2-yl phosphate hydrate 2:1:x, beta-glycerophosphate disodium salt hydrate, ≤1.0 mol % l-alpha-isomer, beta-glycerophosphate disodium salt hydrate, powder, bioreagent, suitable for cell culture, beta-glycerophosphate disodium salt hydrate, ≤2.4 mol % alpha-isomer based on c3h7o6pna2 with x h2o
UQVSFONYLAZJDB-UHFFFAOYSA-N
1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium
131860121
200mM soln.
819-83-0
C3H9Na2O6P
100 mL
Soluble in water.
C(C(CO)OP(=O)(O)O)O.[Na].[Na]
218.052
216.04
Liquid

Chemical Identifiers

819-83-0
218.052
UQVSFONYLAZJDB-UHFFFAOYSA-N
131860121
C(C(CO)OP(=O)(O)O)O.[Na].[Na]
C3H9Na2O6P
MFCD00002135
glycerol 2-phosphate disodium salt hydrate, disodium glycerol 2-phosphate hydrate, beta-glycerophosphoric acid disodium salt, c3h7o6p.2na.h2o, sodium glycerophosphate usan, b-glycerophosphate disodium salt hydrate, sodium 1,3-dihydroxypropan-2-yl phosphate hydrate 2:1:x, beta-glycerophosphate disodium salt hydrate, ≤1.0 mol % l-alpha-isomer, beta-glycerophosphate disodium salt hydrate, powder, bioreagent, suitable for cell culture, beta-glycerophosphate disodium salt hydrate, ≤2.4 mol % alpha-isomer based on c3h7o6pna2 with x h2o
1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only