missing translation for 'onlineSavingsMsg'
Learn More

beta-Methallyl Alcohol 98.0+%, TCI America™

$65.24 - $965.59

Chemical Identifiers

CAS 513-42-8
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00004737
InChI Key BYDRTKVGBRTTIT-UHFFFAOYSA-N
Synonym methallyl alcohol, 2-methyl-2-propen-1-ol, 2-methylallyl alcohol, isopropenyl carbinol, 2-propen-1-ol, 2-methyl, beta-methallyl alcohol, methacryl alcohol, 3-hydroxy-2-methylpropene, beta-methylallyl alcohol, 2-methyl-2-propene-1-ol
PubChem CID 10557
IUPAC Name 2-methylprop-2-en-1-ol
SMILES CC(=C)CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M00915ML
View Documents
TCI America
M00915ML
5 mL N/A
Only null left
 
M009125ML
View Documents
TCI America
M009125ML
25 mL N/A
Only null left
 
M0091500ML
View Documents
TCI America
M0091500ML
500 mL N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

513-42-8
72.107
BYDRTKVGBRTTIT-UHFFFAOYSA-N
10557
CC(=C)CO
C4H8O
MFCD00004737
methallyl alcohol, 2-methyl-2-propen-1-ol, 2-methylallyl alcohol, isopropenyl carbinol, 2-propen-1-ol, 2-methyl, beta-methallyl alcohol, methacryl alcohol, 3-hydroxy-2-methylpropene, beta-methylallyl alcohol, 2-methyl-2-propene-1-ol
2-methylprop-2-en-1-ol

Specifications

513-42-8
113°C
MFCD00004737
2614
BYDRTKVGBRTTIT-UHFFFAOYSA-N
2-methylprop-2-en-1-ol
10557
≥98.0% (GC)
beta-Methallyl Alcohol
Colorless
C4H8O
5 mL
methallyl alcohol, 2-methyl-2-propen-1-ol, 2-methylallyl alcohol, isopropenyl carbinol, 2-propen-1-ol, 2-methyl, beta-methallyl alcohol, methacryl alcohol, 3-hydroxy-2-methylpropene, beta-methylallyl alcohol, 2-methyl-2-propene-1-ol
CC(=C)CO
72.107
72.11
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3872

RTECSNumber : UD5250000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'