Learn More
beta-Propiolactone, 97%
CAS: 57-57-8 | C3H4O2 | 72.06 g/mol
$147.23 - $997.40
Chemical Identifiers
| CAS | 57-57-8 |
|---|---|
| Molecular Formula | C3H4O2 |
| Molecular Weight (g/mol) | 72.06 |
| MDL Number | MFCD00005169 |
| InChI Key | VEZXCJBBBCKRPI-UHFFFAOYSA-N |
| Synonym | beta-propiolactone, propiolactone, 2-oxetanone, propanolide, betaprone, 3-propanolide, 1,3-propiolactone, 3-propiolactone, beta-propionolactone, beta-propiolakton |
| PubChem CID | 2365 |
| ChEBI | CHEBI:49073 |
| IUPAC Name | oxetan-2-one |
| SMILES | O=C1CCO1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2319703
|
Thermo Scientific Chemicals
B2319703 |
1 g |
Each for $147.23
|
|
|||||
|
AAB2319706
|
Thermo Scientific Chemicals
B2319706 |
5 g |
Each for $358.36
|
|
|||||
|
AAB2319714
|
Thermo Scientific Chemicals
B2319714 |
25 g |
Each for $997.40
|
|
|||||
Description
Beta-Propiolactone is widely used as a sterilizing and sporicidal agent, which is used to sterilize blood plasma, vaccines, tissue grafts, surgical instruments and enzymes. It finds an application in the manufacturing of acrylic acid and its esters. It is utilized as an intermediate in the synthesis of synthetic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBeta-Propiolactone is widely used as a sterilizing and sporicidal agent, which is used to sterilize blood plasma, vaccines, tissue grafts, surgical instruments and enzymes. It finds an application in the manufacturing of acrylic acid and its esters. It is utilized as an intermediate in the synthesis of synthetic compounds.
Solubility
Miscible with water and organic solvents.
Notes
Flammable material. Should be stored in a separate safety storage cabinet or room. Keep away from heat and sources of ignition.
Chemical Identifiers
| 57-57-8 | |
| 72.06 | |
| VEZXCJBBBCKRPI-UHFFFAOYSA-N | |
| 2365 | |
| oxetan-2-one |
| C3H4O2 | |
| MFCD00005169 | |
| beta-propiolactone, propiolactone, 2-oxetanone, propanolide, betaprone, 3-propanolide, 1,3-propiolactone, 3-propiolactone, beta-propionolactone, beta-propiolakton | |
| CHEBI:49073 | |
| O=C1CCO1 |
Specifications
| 57-57-8 | |
| 1.146 | |
| 70°C (158°F) | |
| C3H4O2 | |
| MFCD00005169 | |
| UN3382 | |
| 14,7820 | |
| Miscible with water and organic solvents. | |
| O=C1CCO1 | |
| 72.06 | |
| CHEBI:49073 | |
| 97% | |
| beta-Propiolactone |
| -34°C | |
| 161°C to 163°C | |
| 0.97 | |
| 1.413 | |
| 1 g | |
| Moisture sensitive | |
| beta-propiolactone, propiolactone, 2-oxetanone, propanolide, betaprone, 3-propanolide, 1,3-propiolactone, 3-propiolactone, beta-propionolactone, beta-propiolakton | |
| VEZXCJBBBCKRPI-UHFFFAOYSA-N | |
| oxetan-2-one | |
| 2365 | |
| 72.06 | |
| Liquid |
Safety and Handling
GHS H Statement
H330-H350-H227-H315-H319
P201-P202-P210-P235-P260-P264b-P271-P280i-P281-P284-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H330-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, N.O.S.
EINECSNumber : 200-340-1
RTECSNumber : RQ7350000
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only