missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Betaine, 5M Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
$82.70 - $522.63
Chemical Identifiers
CAS | 107-43-7 |
---|---|
Molecular Formula | C5H11NO2 |
Molecular Weight (g/mol) | 117.148 |
MDL Number | MFCD00012123 |
InChI Key | KWIUHFFTVRNATP-UHFFFAOYSA-N |
Synonym | betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin |
PubChem CID | 247 |
ChEBI | CHEBI:17750 |
IUPAC Name | 2-(trimethylazaniumyl)acetate |
SMILES | C[N+](C)(C)CC(=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AAJ77507UCR
|
Thermo Fisher Scientific
J77507UCR |
1.5 mL | Plastic bottle |
Each for $82.70
|
|
||||
AAJ77507VCR
|
Thermo Fisher Scientific
J77507VCR |
5 x 1.5 mL | Plastic bottle |
Case of 5 Each for $174.04
|
|
||||
AAJ77507AB
|
Thermo Fisher Scientific
J77507AB |
10 mL | Plastic bottle |
Each for $183.38
|
|
||||
AAJ77507AE
|
Thermo Fisher Scientific
J77507AE |
100 mL | Plastic bottle |
Each for $522.63
|
|
||||
Chemical Identifiers
107-43-7 | |
117.148 | |
KWIUHFFTVRNATP-UHFFFAOYSA-N | |
247 | |
2-(trimethylazaniumyl)acetate |
C5H11NO2 | |
MFCD00012123 | |
betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin | |
CHEBI:17750 | |
C[N+](C)(C)CC(=O)[O-] |
Specifications
107-43-7 | |
C5H11NO2 | |
1.5 mL | |
betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin | |
C[N+](C)(C)CC(=O)[O-] | |
117.148 | |
CHEBI:17750 | |
Solution |
Plastic bottle | |
MFCD00012123 | |
3537113 | |
KWIUHFFTVRNATP-UHFFFAOYSA-N | |
2-(trimethylazaniumyl)acetate | |
247 | |
Molecular Biology, Ultrapure | |
Betaine |
Safety and Handling
EINECSNumber : 203-490-6
RTECSNumber : DS5900000
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only