missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Betaine Hydrochloride, USP, 98-100.5%, Spectrum™ Chemical
B1104, 590-46-5, C5H11NO2•HCl
Supplier: Spectrum Chemical Mfg Cor B110412KGBL
Description
Spectrum™ Chemical Betaine Hydrochloride, USP is a vitamin-like substance used to treat hypochlorhydria. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
590-46-5 | |
0.1% | |
C5H12ClNO2 | |
12 kg | |
[Cl-].C[N+](C)(C)CC(O)=O | |
153.61 | |
USP |
100% | |
Poly Pail | |
MFCD00011903 | |
HOPSCVCBEOCPJZ-UHFFFAOYSA-N | |
(carboxymethyl)trimethylazanium chloride | |
98 to 100.5% |
Chemical Identifiers
590-46-5 | |
153.61 | |
HOPSCVCBEOCPJZ-UHFFFAOYSA-N | |
[Cl-].C[N+](C)(C)CC(O)=O |
C5H12ClNO2 | |
MFCD00011903 | |
(carboxymethyl)trimethylazanium chloride |